Assay Details
| Gene Symbol: GYPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109960.1 | NP_001103430.1 | 3 - 4 | 255 | 80 | |
| XM_014119221.2 | XP_013974696.1 | 3 - 4 | 295 | 80 | ||
| GenBank mRNA | AB299408.1 | - | 3 - 4 | 255 | 80 | |
More Information
Set Membership: |
|

GYPA
Genomic Map