Assay Details
| Gene Symbol: DEFB4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113715.1 | NP_001107187.1 | 1 - 2 | 133 | 117 | |
| GenBank mRNA | DQ011971.1 | - | 1 - 2 | 133 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001113715.1 transcripts.
Set Membership: |
|

DEFB4A
Genomic Map