Assay Details
| Gene Symbol: BID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251938.1 | NP_001238867.1 | 4 - 5 | 454 | 67 | |
| GenBank mRNA | DN402949.1 | - | 5 - 6 | 462 | 67 | |
| KT693116.1 | - | 3 - 4 | 425 | 67 | ||
More Information
Set Membership: |
|

BID
Genomic Map