Assay Details
| Gene Symbol: GDF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625463.3 | XP_005625520.1 | 2128 | 76 | ||
| XM_005625465.3 | XP_005625522.1 | 2129 | 76 | |||
| XM_014117133.2 | XP_013972608.1 | 2128 | 76 | |||
| XM_014117134.2 | XP_013972609.1 | 2907 | 76 | |||
| XM_022424026.1 | XP_022279734.1 | 956 | 76 | |||
| XM_022424027.1 | XP_022279735.1 | 2129 | 76 | |||
| XM_022424028.1 | XP_022279736.1 | 2479 | 76 | |||
| XM_022424029.1 | XP_022279737.1 | 2733 | 76 | |||
More Information
Set Membership: |
|

GDF11
Genomic Map