Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164487.1 | NP_001157959.1 | 2 - 3 | 344 | 101 | |
| GenBank mRNA | DN397021.1 | - | 2 - 3 | 377 | 101 | |
| DN425872.1 | - | 2 - 3 | 302 | 101 | ||
More Information
Set Membership: |
|

SELENOT
Genomic Map