Assay Details
| Gene Symbol: MIR96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049449.1 | 1 | 44 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-183 | MIR183 | cfa-mir-183 | -51.0 | Sense | Chr. 14 - 7068542 - 7068603 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR96
Genomic Map