Assay Details
| Gene Symbol: CHI3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177807.1 | NP_001171278.1 | 466 | 63 | ||
| XM_005622156.3 | XP_005622213.1 | 1162 | 63 | |||
| GenBank mRNA | GU480918.1 | - | 466 | 63 | ||
More Information
Set Membership: |
|

CHI3L1
Genomic Map