Assay Details
| Gene Symbol: CENPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204929.1 | NP_001191858.1 | 4 - 5 | 271 | 99 | |
| XM_005617935.3 | XP_005617992.1 | 4 - 5 | 377 | 99 | ||
| GenBank mRNA | CF413129.1 | - | 1 - 2 | 546 | 99 | |
More Information
Set Membership: |
|

CENPS
Genomic Map