Assay Details
| Gene Symbol: HMGN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003101.2 | NP_001003101.1 | 1 | 49 | 76 | |
| GenBank mRNA | DN379267.1 | - | 1 | 49 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003101.2 transcripts.
Set Membership: |
|

HMGN2
Genomic Map