Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204899.1 | NP_001191828.1 | 6 - 7 | 496 | 80 | |
| XM_022424914.1 | XP_022280622.1 | 7 - 8 | 577 | 80 | ||
| XM_022424915.1 | XP_022280623.1 | 7 - 8 | 573 | 80 | ||
| XM_022424916.1 | XP_022280624.1 | 7 - 8 | 577 | 80 | ||
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map