Assay Details
| Gene Symbol: RPL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252044.1 | NP_001238973.1 | 1 | 1016 | 109 | |
| XM_005635385.1 | XP_005635442.1 | 1 | 1189 | 109 | ||
| XM_022409578.1 | XP_022265286.1 | 1313 | 109 | |||
| GenBank mRNA | CO589716.1 | - | 1 | 230 | 109 | |
More Information
Set Membership: |
|

RPL21
Genomic Map