Assay Details
| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003254.2 | NP_001003254.1 | 2 - 3 | 188 | 67 | |
| GenBank mRNA | Z49944.1 | - | 1 | 188 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003254.2 | NP_001003254.1 | 2 - 3 | 188 | 67 | |
| GenBank mRNA | Z49944.1 | - | 1 | 188 | 67 | |
Set Membership: |
|