Assay Details
| Gene Symbol: CRYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252397.1 | NP_001239326.1 | 1 - 2 | 113 | 61 | |
| XM_014114419.2 | XP_013969894.1 | 1 - 2 | 139 | 61 | ||
| GenBank mRNA | DN335954.1 | - | 1 - 2 | 102 | 61 | |
More Information
Set Membership: |
|

CRYZ
Genomic Map