Assay Details
| Gene Symbol: MIR592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049506.1 | 1 | 48 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutamate metabotropic receptor 8 | GRM8 | Sense | Chr. 14 - 8982180 - 9719813 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR592
Genomic Map