Assay Details
| Gene Symbol: CEBPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616766.3 | XP_005616823.1 | 1 - 2 | 284 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005616766.3 transcripts.
Set Membership: |
|

CEBPG
Genomic Map