Assay Details
| Gene Symbol: SNHG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038068.1 | 1 - 2 | 54 | 74 | ||
| GenBank mRNA | DR106498.1 | - | 1 - 2 | 118 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sprouty RTK signaling antagonist 4 | SPRY4 | -93.7kb | Sense | Chr. 2 - 37199890 - 37213710 on NCBI Build CanFam3.1 | |
| Downstream | fibroblast growth factor 1 | FGF1 |
ECGF, FGF-1, HBGF-1, aFGF |
158.7kb | Sense | Chr. 2 - 37467431 - 37554770 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

SNHG3
Genomic Map