Assay Details
| Gene Symbol: MIR30D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049243.1 | 1 | 29 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | vacuolar protein sorting 13 homolog B | VPS13B | Anti-sense | Chr. 13 - 1101226 - 1834933 on NCBI Build CanFam3.1 | ||
| Downstream | microRNA mir-599 | MIR599 | cfa-mir-599 | 283.0kb | Sense | Chr. 13 - 1538928 - 1539022 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR30D
Genomic Map