Assay Details
| Gene Symbol: MIR18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049418.1 | 1 | 53 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-17 | MIR17 | cfa-mir-17 | Sense | Chr. 22 - 42478013 - 42478071 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR18A
Genomic Map