Assay Details
| Gene Symbol: MIR149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049473.1 | 1 | 45 | 53 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | spermatogenesis and oogenesis specific basic helix-loop-helix 2 | SOHLH2 | SOSF2 | -281.1kb | Sense | Chr. 25 - 4345785 - 4419352 on NCBI Build CanFam3.1 |
| Downstream | neurobeachin | NBEA | 156.2kb | Anti-sense | Chr. 25 - 4856748 - 5537040 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR149
Genomic Map