Assay Details
| Gene Symbol: SLC46A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120617.1 | 1 - 2 | 1047 | 63 | ||
| GenBank mRNA | AY225149.1 | - | 1 - 2 | 1047 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | post-GPI attachment to proteins GalNAc transferase 4 | PGAP4 |
C11H9orf125, TMEM246 |
-29.1kb | Sense | Chr. 11 - 58197396 - 58218776 on NCBI Build CanFam3.1 |
| Downstream | glutamate ionotropic receptor NMDA type subunit 3A | GRIN3A | 21.2kb | Sense | Chr. 11 - 58271808 - 58421887 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

SLC46A2
Genomic Map