Assay Details
| Gene Symbol: MIR19B-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049400.1 | 1 | 29 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glypican 3 | GPC3 | -49.2kb | Sense | Chr. X - 104313426 - 104751215 on NCBI Build CanFam3.1 | |
| Downstream |
nuclease EXOG, mitochondrial-like |
LOC100686111 | 56.6kb | Anti-sense | Chr. X - 104857047 - 104858004 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR19B-2
Genomic Map