Assay Details
| Gene Symbol: MIRLET7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049422.1 | 1 | 39 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | retrovirus-related Pol polyprotein | LOC106559347 | -187.5kb | Anti-sense | Chr. 10 - 3279093 - 3287181 on NCBI Build CanFam3.1 | |
| Downstream | TAFA chemokine like family member 2 | TAFA2 | FAM19A2 | 1.5mb | Sense | Chr. 10 - 4998538 - 5134970 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIRLET7B
Genomic Map