Assay Details
| Gene Symbol: MIR2483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128883.1 | 1 | 83 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | teneurin transmembrane protein 1 | TENM1 | ODZ1 | Sense | Chr. X - 95766173 - 96565995 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR2483
Genomic Map