Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284448.1 | NP_001271377.1 | 3 - 4 | 354 | 61 | |
| GenBank mRNA | JN790861.1 | - | 3 - 4 | 354 | 61 | |
More Information
Set Membership: |
|

NCR1
Genomic Map