Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302665.1 | NP_001289594.1 | 3 - 4 | 659 | 50 | |
| GenBank mRNA | DN379206.1 | - | 2 - 3 | 414 | 50 | |
More Information
Set Membership: |
|

SLAMF9
Genomic Map