Assay Details
| Gene Symbol: TIMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314106.1 | NP_001301035.1 | 267 | 125 | ||
| GenBank mRNA | LC041119.1 | - | 267 | 125 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TIMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314106.1 | NP_001301035.1 | 267 | 125 | ||
| GenBank mRNA | LC041119.1 | - | 267 | 125 | ||
Set Membership: |
|