Assay Details
| Gene Symbol: Psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500886.2 | XP_003500934.1 | 1 - 2 | 227 | 90 | |
| GenBank mRNA | JI870400.1 | - | 1 - 2 | 197 | 90 | |
| JP055337.1 | - | 1 - 2 | 197 | 90 | ||
More Information
Set Membership: |
|

Psmb2
Genomic Map