Assay Details
| Gene Symbol: Desi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514730.2 | XP_003514778.1 | 2 - 3 | 353 | 84 | |
| XM_007654786.1 | XP_007652976.1 | 2 - 3 | 355 | 84 | ||
| GenBank mRNA | JI866689.1 | - | 2 - 3 | 204 | 84 | |
| JP066092.1 | - | 2 - 3 | 204 | 84 | ||
More Information
Set Membership: |
|

Desi1
Genomic Map