Assay Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496243.2 | XP_003496291.1 | 5 - 6 | 1008 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou6f2| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496243.2 | XP_003496291.1 | 5 - 6 | 1008 | 88 | |
Set Membership: |
|