Assay Details
| Gene Symbol: Ap2m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509612.2 | XP_003509660.1 | 10 - 11 | 1223 | 66 | |
| XM_003509613.2 | XP_003509661.1 | 9 - 10 | 1218 | 66 | ||
| GenBank mRNA | JI873823.1 | - | 10 - 11 | 1205 | 66 | |
| JP038718.1 | - | 10 - 11 | 1205 | 66 | ||
More Information
Set Membership: |
|

Ap2m1
Genomic Map