Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 83 - 84 | 20020 | 72 | |
| XM_007645556.1 | XP_007643746.1 | 83 - 84 | 20020 | 72 | ||
| XM_007645557.1 | XP_007643747.1 | 82 - 83 | 19957 | 72 | ||
| XM_007645558.1 | XP_007643748.1 | 78 - 79 | 13924 | 72 | ||
| XM_007645559.1 | XP_007643749.1 | 78 - 79 | 13924 | 72 | ||
| XM_007645560.1 | XP_007643750.1 | 77 - 78 | 13861 | 72 | ||
| XM_007645561.1 | XP_007643751.1 | 27 - 28 | 8648 | 72 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map