Assay Details
Gene Symbol: Rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641392.1 | XP_007639582.1 | 4 - 5 | 676 | 82 | |
GenBank mRNA | JI864868.1 | - | 1 | 676 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
