Assay Details
| Gene Symbol: Cdc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644222.1 | XP_007642412.1 | 4 - 5 | 416 | 74 | |
| GenBank mRNA | JI867963.1 | - | 4 - 5 | 398 | 74 | |
| JP045669.1 | - | 4 - 5 | 398 | 74 | ||
More Information
Set Membership: |
|

Cdc42
Genomic Map