Assay Details
| Gene Symbol: Wbscr22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500922.2 | XP_003500970.1 | 2 - 3 | 134 | 69 | |
| GenBank mRNA | JP044840.1 | - | 2 - 3 | 136 | 69 | |
More Information
Set Membership: |
|

Wbscr22
Genomic Map