Assay Details
| Gene Symbol: Large | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505031.2 | XP_003505079.1 | 14 - 15 | 2183 | 60 | |
| XM_007645529.1 | XP_007643719.1 | 16 - 17 | 2696 | 60 | ||
| XM_007645530.1 | XP_007643720.1 | 16 - 17 | 2693 | 60 | ||
| XM_007645531.1 | XP_007643721.1 | 14 - 15 | 2259 | 60 | ||
| XM_007645532.1 | XP_007643722.1 | 16 - 17 | 2603 | 60 | ||
| XM_007645534.1 | XP_007643724.1 | 16 - 17 | 2695 | 60 | ||
| XM_007645535.1 | XP_007643725.1 | 16 - 17 | 2554 | 60 | ||
| XM_007645536.1 | XP_007643726.1 | 14 - 15 | 2455 | 60 | ||
| XM_007645537.1 | XP_007643727.1 | 15 - 16 | 2601 | 60 | ||
More Information
Set Membership: |
|

Large
Genomic Map