Assay Details
| Gene Symbol: Nubp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642189.1 | XP_007640379.1 | 3 - 4 | 512 | 58 | |
| XM_007642190.1 | XP_007640380.1 | 3 - 4 | 393 | 58 | ||
| XM_007642191.1 | XP_007640381.1 | 3 - 4 | 516 | 58 | ||
More Information
Set Membership: |
|

Nubp2
Genomic Map