Assay Details
| Gene Symbol: Cfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651897.1 | XP_007650087.1 | 1 - 2 | 365 | 87 | |
| GenBank mRNA | JP037362.1 | - | 1 - 2 | 365 | 87 | |
More Information
Set Membership: |
|

Cfl2
Genomic Map