Assay Details
| Gene Symbol: Vdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498509.2 | XP_003498557.2 | 1 | 396 | 83 | |
| GenBank mRNA | JI877718.1 | - | 1 | 396 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vdac1| Gene Symbol: Vdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498509.2 | XP_003498557.2 | 1 | 396 | 83 | |
| GenBank mRNA | JI877718.1 | - | 1 | 396 | 83 | |
Set Membership: |
|