Assay Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642220.1 | XP_007640410.1 | 15 - 16 | 3044 | 66 | |
| GenBank mRNA | JI884108.1 | - | 2 - 3 | 134 | 66 | |
More Information
Set Membership: |
|

Cramp1l
Genomic Map