Assay Details
| Gene Symbol: Eri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497390.2 | XP_003497438.1 | 3 - 4 | 488 | 74 | |
| XM_007611506.1 | XP_007609696.1 | 3 - 4 | 474 | 74 | ||
| XM_007611516.1 | XP_007609706.1 | 2 - 3 | 178 | 74 | ||
| XM_007611520.1 | XP_007609710.1 | 4 - 5 | 496 | 74 | ||
| GenBank mRNA | JP053053.1 | - | 3 - 4 | 487 | 74 | |
More Information
Set Membership: |
|

Eri3
Genomic Map