Assay Details
| Gene Symbol: Bzw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507033.2 | XP_003507081.1 | 9 - 10 | 1126 | 69 | |
| XM_007647582.1 | XP_007645772.1 | 9 - 10 | 1182 | 69 | ||
| GenBank mRNA | JP039267.1 | - | 9 - 10 | 1209 | 69 | |
More Information
Set Membership: |
|

Bzw1
Genomic Map