Assay Details
| Gene Symbol: Layn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246681.1 | NP_001233610.1 | 1 - 2 | 205 | 65 | |
| GenBank mRNA | AF093673.1 | - | 1 - 2 | 229 | 65 | |
| JI889967.1 | - | 1 - 2 | 209 | 65 | ||
| JP045844.1 | - | 1 - 2 | 209 | 65 | ||
More Information
Set Membership: |
|

Layn
Genomic Map