Assay Details
| Gene Symbol: Neu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246735.2 | NP_001233664.1 | 3 - 4 | 392 | 104 | |
| GenBank mRNA | U06143.1 | - | 3 - 4 | 392 | 104 | |
More Information
Set Membership: |
|

Neu2
Genomic Map