Assay Details
| Gene Symbol: Slc19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246831.1 | NP_001233760.1 | 2 - 3 | 287 | 60 | |
| XM_007641311.1 | XP_007639501.1 | 3 - 4 | 439 | 60 | ||
| XM_007641312.1 | XP_007639502.1 | 3 - 4 | 409 | 60 | ||
| XM_007641313.1 | XP_007639503.1 | 3 - 4 | 481 | 60 | ||
| XM_007641314.1 | XP_007639504.1 | 3 - 4 | 377 | 60 | ||
| XM_007641315.1 | XP_007639505.1 | 2 - 3 | 292 | 60 | ||
| XM_007641316.1 | XP_007639506.1 | 2 - 3 | 266 | 60 | ||
| GenBank mRNA | JI863119.1 | - | 3 - 4 | 380 | 60 | |
| JP038402.1 | - | 3 - 4 | 380 | 60 | ||
| U03031.1 | - | 2 - 3 | 287 | 60 | ||
More Information
Set Membership: |
|

Slc19a1
Genomic Map