Assay Details
| Gene Symbol: Cbr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244007.1 | NP_001230936.1 | 1 - 2 | 399 | 87 | |
| GenBank mRNA | AB020238.1 | - | 1 - 2 | 399 | 87 | |
| AB043543.3 | - | 1 - 2 | 399 | 87 | ||
More Information
Set Membership: |
|

Cbr3
Genomic Map