Assay Details
| Gene Symbol: Sepw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256880.1 | NP_001243809.1 | 1 | 296 | 82 | |
| GenBank mRNA | JI872664.1 | - | 1 | 297 | 82 | |
| JP041795.1 | - | 1 | 296 | 82 | ||
More Information
Set Membership: |
|

Sepw1
Genomic Map