Assay Details
| Gene Symbol: Sec22b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246746.1 | NP_001233675.1 | 1 - 2 | 160 | 63 | |
| GenBank mRNA | JI875226.1 | - | 1 - 2 | 215 | 63 | |
| JP060503.1 | - | 1 - 2 | 215 | 63 | ||
| U91742.1 | - | 1 - 2 | 160 | 63 | ||
More Information
Set Membership: |
|

Sec22b
Genomic Map