Assay Details
| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246793.1 | NP_001233722.1 | 2 - 3 | 483 | 69 | |
| GenBank mRNA | DQ981502.1 | - | 2 - 3 | 483 | 69 | |
More Information
Set Membership: |
|

Dph2
Genomic Map