Assay Details
| Gene Symbol: Adsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244045.1 | NP_001230974.1 | 9 - 10 | 1031 | 60 | |
| GenBank mRNA | GQ984234.1 | - | 9 - 10 | 1031 | 60 | |
| GQ984235.1 | - | 9 - 10 | 1029 | 60 | ||
| JI874073.1 | - | 9 - 10 | 1037 | 60 | ||
| JP065740.1 | - | 9 - 10 | 1037 | 60 | ||
More Information
Set Membership: |
|

Adsl
Genomic Map