Assay Details
| Gene Symbol: Ap3m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244405.1 | NP_001231334.1 | 4 - 5 | 672 | 65 | |
| XM_007652612.1 | XP_007650802.1 | 5 - 6 | 709 | 65 | ||
| XM_007652618.1 | XP_007650808.1 | 5 - 6 | 839 | 65 | ||
| XM_007652623.1 | XP_007650813.1 | 4 - 5 | 573 | 65 | ||
| GenBank mRNA | AY346109.1 | - | 5 - 6 | 939 | 65 | |
| JI885336.1 | - | 5 - 6 | 1030 | 65 | ||
| JP040800.1 | - | 5 - 6 | 712 | 65 | ||
More Information
Set Membership: |
|

Ap3m1
Genomic Map